Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAATGGTCATAGTCAAAAACTG[A/G]CCACTCCCCTGCTCAAGAGTCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCFD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCFD2 - sec1 family domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152540.3 | Intron | NP_689753.2 | ||||
XM_011534375.2 | Intron | XP_011532677.1 | ||||
XM_011534376.2 | Intron | XP_011532678.1 | ||||
XM_011534378.2 | Intron | XP_011532680.1 | ||||
XM_017007786.1 | Intron | XP_016863275.1 | ||||
XM_017007787.1 | Intron | XP_016863276.1 |