Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCGGGAAAGGCAGGGGGAAGAA[A/G]AGGAGGTACCCCAGCCTCACCAACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610203 MIM: 614632 | ||||||||||||||||||||
Literature Links: |
CRIPAK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRIPAK - cysteine rich PAK1 inhibitor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UVSSA - UV stimulated scaffold protein A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317934.1 | 2117 | Silent Mutation | AAA,AAG | K,K 652 | NP_001304863.1 | |
NM_001317935.1 | 2117 | Silent Mutation | AAA,AAG | K,K 652 | NP_001304864.1 | |
NM_020894.3 | 2117 | Silent Mutation | AAA,AAG | K,K 652 | NP_065945.2 | |
XM_017008490.1 | 2117 | Silent Mutation | AAA,AAG | K,K 652 | XP_016863979.1 | |
XM_017008491.1 | 2117 | Silent Mutation | AAA,AAG | K,K 652 | XP_016863980.1 | |
XM_017008492.1 | 2117 | Silent Mutation | AAA,AAG | K,K 652 | XP_016863981.1 | |
XM_017008493.1 | 2117 | Silent Mutation | AAA,AAG | K,K 652 | XP_016863982.1 | |
XM_017008494.1 | 2117 | Silent Mutation | AAA,AAG | K,K 652 | XP_016863983.1 | |
XM_017008495.1 | 2117 | Silent Mutation | AAA,AAG | K,K 556 | XP_016863984.1 | |
XM_017008496.1 | 2117 | Intron | XP_016863985.1 | |||
XM_017008497.1 | 2117 | Intron | XP_016863986.1 | |||
XM_017008498.1 | 2117 | Intron | XP_016863987.1 | |||
XM_017008499.1 | 2117 | Intron | XP_016863988.1 | |||
XM_017008500.1 | 2117 | Silent Mutation | AAA,AAG | K,K 262 | XP_016863989.1 |