Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGTTCTTCTTCTAGACACTGAA[A/G]ATGTTTCAGTTCTGTGGCCTAGAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147680 | ||||||||||||||||||||
Literature Links: |
IL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL2 - interleukin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000586.3 | 281 | Missense Mutation | CTT,TTT | L,F 76 | NP_000577.2 | |
XM_017008177.1 | 281 | Missense Mutation | CTT,TTT | L,F 76 | XP_016863666.1 |