Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCAGGGGCCTGGGTCTGCGGGGC[C/T]ACCATGGCAACCGGGGGTGTCTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606026 MIM: 602952 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR943 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR943 - microRNA 943 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NELFA - negative elongation factor complex member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005663.4 | 1521 | Silent Mutation | GTA,GTG | V,V 426 | NP_005654.3 | |
XM_017008589.1 | 1521 | Silent Mutation | GTA,GTG | V,V 454 | XP_016864078.1 |
SCARNA22 - small Cajal body-specific RNA 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WHSC1 - Wolf-Hirschhorn syndrome candidate 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |