Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAATGGAATACAGTAAGGATAAT[A/G]ATAACTTGGTTTCGATCTCAGTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608301 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LGI2 - leucine rich repeat LGI family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018176.3 | 5992 | UTR 3 | NP_060646.2 | |||
XM_011513850.2 | 5992 | Intron | XP_011512152.1 | |||
XM_017008356.1 | 5992 | Intron | XP_016863845.1 |