Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGCTGCTTGGAGCTTTGACAGT[A/T]GTGAAGGAACTAGAGTTTTGATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PP12613 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PP12613 - uncharacterized LOC100192379 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM155 - transmembrane protein 155 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317837.1 | 1109 | UTR 3 | NP_001304766.1 | |||
NM_001317838.1 | 1109 | UTR 3 | NP_001304767.1 | |||
NM_001317839.1 | 1109 | UTR 3 | NP_001304768.1 | |||
NM_001317841.1 | 1109 | UTR 3 | NP_001304770.1 | |||
NM_001317842.1 | 1109 | UTR 3 | NP_001304771.1 | |||
NM_152399.3 | 1109 | UTR 3 | NP_689612.3 | |||
XM_005262737.2 | 1109 | UTR 3 | XP_005262794.1 | |||
XM_017007725.1 | 1109 | UTR 3 | XP_016863214.1 | |||
XM_017007726.1 | 1109 | UTR 3 | XP_016863215.1 |