Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAATCACGGTGCTTTATACTTA[C/G]TTTTAATTTCTGCACTGAAAAAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604734 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR3138 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR3138 - microRNA 3138 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR1 - WD repeat domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005112.4 | 3182 | UTR 3 | NP_005103.2 | |||
NM_017491.3 | 3182 | UTR 3 | NP_059830.1 | |||
XM_017008880.1 | 3182 | UTR 3 | XP_016864369.1 |