Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTATTCAGGCTGTTGAGACTGGAG[C/T]CCGGGACGCCTGTCACTGCTGAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601542 | ||||||||||||||||||||
Literature Links: |
PITX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PITX2 - paired like homeodomain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000325.5 | 2158 | Missense Mutation | GAC,GGC | D,G 243 | NP_000316.2 | |
NM_001204397.1 | 2158 | Missense Mutation | GAC,GGC | D,G 236 | NP_001191326.1 | |
NM_001204398.1 | 2158 | Missense Mutation | GAC,GGC | D,G 236 | NP_001191327.1 | |
NM_001204399.1 | 2158 | Missense Mutation | GAC,GGC | D,G 190 | NP_001191328.1 | |
NM_153426.2 | 2158 | Missense Mutation | GAC,GGC | D,G 236 | NP_700475.1 | |
NM_153427.2 | 2158 | Missense Mutation | GAC,GGC | D,G 190 | NP_700476.1 | |
XM_011532027.2 | 2158 | Missense Mutation | GAC,GGC | D,G 190 | XP_011530329.1 |