Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATGTGCATAAATTCAAGCTTGT[C/T]GGCATACTGTTTCAGCATGGCACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600636 | ||||||||||||||||||||
Literature Links: |
CASP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CASP3 - caspase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004346.3 | 945 | Missense Mutation | AAC,GAC | N,D 228 | NP_004337.2 | |
NM_032991.2 | 945 | Missense Mutation | AAC,GAC | N,D 228 | NP_116786.1 | |
XM_011532301.1 | 945 | Missense Mutation | AAC,GAC | N,D 228 | XP_011530603.1 |
LVCAT8 - liver cancer-associated transcript 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |