Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTAGTGCTGGCGATCCTGAACA[A/G]GGTGGCACCGCAGGCGTGCCCGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603746 | ||||||||||||||||||||
Literature Links: |
SLIT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLIT2 - slit guidance ligand 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289135.2 | 1976 | Missense Mutation | AAG,AGG | K,R 22 | NP_001276064.1 | |
NM_001289136.2 | 1976 | Missense Mutation | AAG,AGG | K,R 22 | NP_001276065.1 | |
NM_004787.3 | 1976 | Missense Mutation | AAG,AGG | K,R 22 | NP_004778.1 | |
XM_005248211.3 | 1976 | Missense Mutation | AAG,AGG | K,R 22 | XP_005248268.1 | |
XM_006713986.3 | 1976 | Missense Mutation | AAG,AGG | K,R 22 | XP_006714049.1 | |
XM_011513909.2 | 1976 | Intron | XP_011512211.1 | |||
XM_011513910.2 | 1976 | Intron | XP_011512212.2 | |||
XM_017008845.1 | 1976 | Intron | XP_016864334.1 |