Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGCCTTCAGAAATAGCTTCCAC[A/G]ACTTGCAAATTTGACTTATTTTCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600058 | ||||||||||||||||||||
Literature Links: |
TXK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TXK - TXK tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003328.2 | 1511 | Silent Mutation | GTC,GTT | V,V 475 | NP_003319.2 | |
XM_011513747.2 | 1511 | Silent Mutation | GTC,GTT | V,V 456 | XP_011512049.1 | |
XM_011513748.2 | 1511 | Silent Mutation | GTC,GTT | V,V 237 | XP_011512050.1 | |
XM_017008580.1 | 1511 | Silent Mutation | GTC,GTT | V,V 478 | XP_016864069.1 | |
XM_017008581.1 | 1511 | Silent Mutation | GTC,GTT | V,V 450 | XP_016864070.1 | |
XM_017008582.1 | 1511 | Silent Mutation | GTC,GTT | V,V 286 | XP_016864071.1 |