Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGCACTGCACTCCGCTGATTTT[C/T]CCCGGGGAAGGGAAAAAGGTGGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HELT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HELT - helt bHLH transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300781.1 | 299 | Missense Mutation | CCC,TCC | P,S 71 | NP_001287710.1 | |
NM_001300782.1 | 299 | Missense Mutation | CCC,TCC | P,S 71 | NP_001287711.1 | |
XM_017008186.1 | 299 | Missense Mutation | CCC,TCC | P,S 33 | XP_016863675.1 |