Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTTGTGCATTCCACCATGAGCT[A/G]GAGAAAGCCATCTGCAGGTAAATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614047 | ||||||||||||||||||||
Literature Links: |
EPGN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPGN - epithelial mitogen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270989.1 | 301 | Silent Mutation | CTA,CTG | L,L 79 | NP_001257918.1 | |
NM_001270990.1 | 301 | Silent Mutation | CTA,CTG | L,L 79 | NP_001257919.1 | |
NM_001270991.1 | 301 | Silent Mutation | CTA,CTG | L,L 70 | NP_001257920.1 | |
NM_001270992.1 | 301 | Silent Mutation | CTA,CTG | L,L 79 | NP_001257921.1 | |
NM_001270993.1 | 301 | Silent Mutation | CTA,CTG | L,L 70 | NP_001257922.1 | |
XM_005265666.4 | 301 | Silent Mutation | CTA,CTG | L,L 70 | XP_005265723.1 | |
XM_011531817.2 | 301 | Silent Mutation | CTA,CTG | L,L 70 | XP_011530119.1 |
LOC105377276 - uncharacterized LOC105377276 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTHFD2L - methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |