Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGACAACTGGGAAATGGTGGAACA[C/T]GTGACCAGCTGATGCCGCTTCCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608242 | ||||||||||||||||||||
Literature Links: |
HERC5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HERC5 - HECT and RLD domain containing E3 ubiquitin protein ligase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016323.3 | 1666 | Missense Mutation | CGT,TGT | R,C 334 | NP_057407.2 | |
XM_011532022.2 | 1666 | Missense Mutation | CGT,TGT | R,C 485 | XP_011530324.2 |