Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTGGGGGAGAGATGCTGTGCGG[C/T]GGCTTCTACCCTCGGCTGTCCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606178 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HHIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HHIP - hedgehog interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022475.2 | 759 | Silent Mutation | GGC,GGT | G,G 70 | NP_071920.1 | |
XM_005263178.4 | 759 | Silent Mutation | GGC,GGT | G,G 70 | XP_005263235.1 | |
XM_006714288.3 | 759 | Silent Mutation | GGC,GGT | G,G 70 | XP_006714351.1 |
HHIP-AS1 - HHIP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |