Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCACCTGGACGAGTCCCTCCAGA[A/C]AAAGGCACTGCTCAACGAGGAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604863 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRAT - lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301645.1 | 457 | Missense Mutation | AAA,ACA | K,T 133 | NP_001288574.1 | |
NM_004744.4 | 457 | Missense Mutation | AAA,ACA | K,T 133 | NP_004735.2 |