Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAAAAATGAATTATCCTCCTTC[A/G]AACTATGTCATGAACTTGAAGTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608252 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AFAP1 - actin filament associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134647.1 | 6422 | UTR 3 | NP_001128119.1 | |||
NM_198595.2 | 6422 | UTR 3 | NP_940997.1 | |||
XM_006713909.3 | 6422 | UTR 3 | XP_006713972.1 | |||
XM_011513544.2 | 6422 | UTR 3 | XP_011511846.1 | |||
XM_017008535.1 | 6422 | UTR 3 | XP_016864024.1 | |||
XM_017008536.1 | 6422 | Intron | XP_016864025.1 |
AFAP1-AS1 - AFAP1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |