Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTTAGCATTTTTTTATGAATGC[A/G]TTCATAATCCAATTCCTGTAAAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603370 | ||||||||||||||||||||
Literature Links: |
UGDH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UGDH - UDP-glucose 6-dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184700.1 | 1602 | Missense Mutation | CGC,TGC | R,C 360 | NP_001171629.1 | |
NM_001184701.1 | 1602 | Missense Mutation | CGC,TGC | R,C 330 | NP_001171630.1 | |
NM_003359.3 | 1602 | Missense Mutation | CGC,TGC | R,C 427 | NP_003350.1 | |
XM_005262667.2 | 1602 | Missense Mutation | CGC,TGC | R,C 440 | XP_005262724.1 |