Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAACACTTACCAAGTAACTCAG[A/G]CACACTCACCGCCTTTTGCTGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616693 | ||||||||||||||||||||
Literature Links: |
ASIC5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASIC5 - acid sensing ion channel subunit family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017419.2 | 1362 | Missense Mutation | CCT,TCT | P,S 439 | NP_059115.1 | |
XM_017008291.1 | 1362 | Missense Mutation | CCT,TCT | P,S 397 | XP_016863780.1 |