Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGGAGCCATGAAACCTTCAGGT[C/G]GTTTCTGGCCTTCTCGAGCAAGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603262 | ||||||||||||||||||||
Literature Links: |
PAPSS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAPSS1 - 3'-phosphoadenosine 5'-phosphosulfate synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005443.4 | 1952 | Missense Mutation | CCA,CGA | P,R 601 | NP_005434.4 | |
XM_011532400.1 | 1952 | Missense Mutation | CCA,CGA | P,R 580 | XP_011530702.1 | |
XM_011532401.1 | 1952 | Missense Mutation | CCA,CGA | P,R 580 | XP_011530703.1 |