Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCACCTTGGTGGGTGTGCCAGAG[C/T]ACACATCCTGGAAAACAGAAGCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602422 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM53A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM53A - family with sequence similarity 53 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLBP - stem-loop binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306074.1 | 1257 | Missense Mutation | TAC,TGC | Y,C 200 | NP_001293003.1 | |
NM_001306075.1 | 1257 | Missense Mutation | TAC,TGC | Y,C 196 | NP_001293004.1 | |
NM_006527.3 | 1257 | Missense Mutation | TAC,TGC | Y,C 235 | NP_006518.1 |