Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATAGAGCTCACCCGCTCCCTGTC[C/G]GACAACGTGAACCTGCCCCAGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613166 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCLK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCLK2 - doublecortin like kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040260.3 | 1087 | Silent Mutation | TCC,TCG | S,S 111 | NP_001035350.2 | |
NM_001040261.4 | 1087 | Silent Mutation | TCC,TCG | S,S 111 | NP_001035351.4 | |
XM_005262785.2 | 1087 | Silent Mutation | TCC,TCG | S,S 111 | XP_005262842.1 | |
XM_005262786.2 | 1087 | Silent Mutation | TCC,TCG | S,S 111 | XP_005262843.1 | |
XM_017007832.1 | 1087 | Silent Mutation | TCC,TCG | S,S 111 | XP_016863321.1 | |
XM_017007833.1 | 1087 | Silent Mutation | TCC,TCG | S,S 111 | XP_016863322.1 |