Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCCCACAGTGGCCTCCGCACAG[C/T]CCCCAGGACGACCACAGGGAGACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605918 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPON2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPON2 - spondin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128325.2 | 1190 | Silent Mutation | GGA,GGG | G,G 287 | NP_001121797.1 | |
NM_001199021.1 | 1190 | Silent Mutation | GGA,GGG | G,G 287 | NP_001185950.1 | |
NM_012445.3 | 1190 | Silent Mutation | GGA,GGG | G,G 287 | NP_036577.1 |