Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCCTCTACTGCAGAGTGGGCAT[C/G]GGTTTCCATCTGCAGATCTACCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 165190 | ||||||||||||||||||||
Literature Links: |
FGF5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FGF5 - fibroblast growth factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291812.1 | 528 | Intron | NP_001278741.1 | |||
NM_004464.3 | 528 | Missense Mutation | ATC,ATG | I,M 97 | NP_004455.2 | |
NM_033143.2 | 528 | Missense Mutation | ATC,ATG | I,M 97 | NP_149134.1 |