Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGGATCCATAGGTGGATATTTT[C/T]GGCCTTTGCTTTTTCCAAGGCATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615039 | ||||||||||||||||||||
Literature Links: |
NDST4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDST4 - N-deacetylase and N-sulfotransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022569.2 | 2151 | Missense Mutation | CAA,CGA | Q,R 825 | NP_072091.1 | |
XM_017008545.1 | 2151 | Missense Mutation | CAA,CGA | Q,R 446 | XP_016864034.1 | |
XM_017008546.1 | 2151 | Missense Mutation | CAA,CGA | Q,R 446 | XP_016864035.1 |