Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCGTTTTAAAACTGCCAGGAAAC[A/G]CTCAGGAAGGATCCTAAAAGAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612831 | ||||||||||||||||||||
Literature Links: |
HSD17B11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSD17B11 - hydroxysteroid 17-beta dehydrogenase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016245.4 | 1059 | Missense Mutation | CGT,TGT | R,C 276 | NP_057329.3 | |
XM_011532021.1 | 1059 | Intron | XP_011530323.1 |