Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCATCTGGTCTTGAGTGAGCTT[C/G]TGTGCTTTGCTGACCCGGGTGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614532 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASGEF1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RASGEF1B - RasGEF domain family member 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300735.1 | 1727 | UTR 3 | NP_001287664.1 | |||
NM_001300736.1 | 1727 | UTR 3 | NP_001287665.1 | |||
NM_152545.2 | 1727 | UTR 3 | NP_689758.1 | |||
XM_017007812.1 | 1727 | UTR 3 | XP_016863301.1 | |||
XM_017007813.1 | 1727 | UTR 3 | XP_016863302.1 |