Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTAGTCACATAATCTTCCTCCAC[A/G]CTCTTTTCTACCTTTTGTAATAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603296 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJB14 - DnaJ heat shock protein family (Hsp40) member B14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031723.3 | 1496 | Silent Mutation | NP_001026893.1 | |||
NM_001278310.1 | 1496 | Silent Mutation | NP_001265239.1 | |||
NM_001278311.1 | 1496 | Intron | NP_001265240.1 | |||
XM_011532262.2 | 1496 | Silent Mutation | XP_011530564.1 | |||
XM_011532263.2 | 1496 | Missense Mutation | XP_011530565.1 | |||
XM_011532265.2 | 1496 | Missense Mutation | XP_011530567.1 | |||
XM_017008628.1 | 1496 | Missense Mutation | XP_016864117.1 |
LAMTOR3 - late endosomal/lysosomal adaptor, MAPK and MTOR activator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |