Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTATAATTTGGACTTGGGAAAC[A/C]GGCATTGTTTCCTTTTTCAAAGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TECRL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TECRL - trans-2,3-enoyl-CoA reductase-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010874.4 | 957 | Missense Mutation | GGT,TGT | G,C 282 | NP_001010874.2 | |
XM_005265662.4 | 957 | Missense Mutation | GGT,TGT | G,C 267 | XP_005265719.1 | |
XM_005265663.2 | 957 | Missense Mutation | GGT,TGT | G,C 282 | XP_005265720.1 | |
XM_005265664.2 | 957 | Missense Mutation | GGT,TGT | G,C 282 | XP_005265721.1 | |
XM_005265665.3 | 957 | Missense Mutation | GGT,TGT | G,C 282 | XP_005265722.1 | |
XM_017007959.1 | 957 | Intron | XP_016863448.1 | |||
XM_017007960.1 | 957 | Intron | XP_016863449.1 |