Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAGAACAAGATCAATGCAATAAC[C/T]ATCCAGGCTACCAAGATCATTGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SMIM14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SMIM14 - small integral membrane protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317896.1 | 368 | Missense Mutation | ATA,ATG | I,M 61 | NP_001304825.1 | |
NM_001317897.1 | 368 | Missense Mutation | ATA,ATG | I,M 61 | NP_001304826.1 | |
NM_174921.2 | 368 | Missense Mutation | ATA,ATG | I,M 61 | NP_777581.1 | |
XM_011513640.2 | 368 | Missense Mutation | ATA,ATG | I,M 84 | XP_011511942.1 | |
XM_017007862.1 | 368 | Missense Mutation | ATA,ATG | I,M 61 | XP_016863351.1 | |
XM_017007863.1 | 368 | Missense Mutation | ATA,ATG | I,M 61 | XP_016863352.1 |
UGDH-AS1 - UGDH antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |