Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACACGTGCCCATGTGGAGTGCC[C/T]GCCTGCTCACACAAAGCCCTGGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610203 MIM: 614632 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRIPAK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRIPAK - cysteine rich PAK1 inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175918.3 | 4142 | Silent Mutation | CCC,CCT | P,P 394 | NP_787114.2 |
NKX1-1 - NK1 homeobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UVSSA - UV stimulated scaffold protein A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |