Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGATGATAATTTGGATTTGTCA[G/T]TATTCCATTTCTCTTCTTCTGTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ANKRD50 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD50 - ankyrin repeat domain 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167882.1 | 5855 | Missense Mutation | ATG,CTG | M,L 1183 | NP_001161354.1 | |
NM_020337.2 | 5855 | Missense Mutation | ATG,CTG | M,L 1362 | NP_065070.1 | |
XM_017008471.1 | 5855 | Missense Mutation | ATG,CTG | M,L 1362 | XP_016863960.1 |