Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTACTTTGGGCAGCAGAGCAATT[C/T]GCTGAAGTTGCAGTAACAGATCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603163 | ||||||||||||||||||||
Literature Links: |
SCRG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCRG1 - stimulator of chondrogenesis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011531578.1 | 349 | Missense Mutation | AAA,GAA | K,E 114 | XP_011529880.1 | |
XM_017007678.1 | 349 | Missense Mutation | AAA,GAA | K,E 112 | XP_016863167.1 |