Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTTAATCCAAATAATGTCCACC[A/G]TCCTCCAGGCTTCTCACAAACAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605236 | ||||||||||||||||||||
Literature Links: |
ATP10D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP10D - ATPase phospholipid transporting 10D (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CORIN - corin, serine peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278585.1 | 3146 | Missense Mutation | CGG,TGG | R,W 893 | NP_001265514.1 | |
NM_001278586.1 | 3146 | Intron | NP_001265515.1 | |||
NM_006587.3 | 3146 | Missense Mutation | CGG,TGG | R,W 997 | NP_006578.2 |