Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCATCGGCTTGGTTGCCCGTTTGC[A/G]GTCAGTGGCCAGCCCCAGCCAGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00575 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00575 - long intergenic non-protein coding RNA 575 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCD5 - stearoyl-CoA desaturase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037582.2 | 1248 | Missense Mutation | CGC,TGC | R,C 310 | NP_001032671.2 | |
NM_024906.2 | 1248 | Intron | NP_079182.2 |