Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAACTCTGGCCACCAGGATGCCC[A/G]CTCCAACACCTTGCTTTCCTGCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603402 | ||||||||||||||||||||
Literature Links: |
ACOX3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACOX3 - acyl-CoA oxidase 3, pristanoyl | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101667.1 | 2059 | UTR 3 | NP_001095137.1 | |||
NM_003501.2 | 2059 | Missense Mutation | CGG,TGG | R,W 679 | NP_003492.2 | |
XM_005248011.4 | 2059 | Missense Mutation | CGG,TGG | R,W 679 | XP_005248068.1 | |
XM_005248013.3 | 2059 | UTR 3 | XP_005248070.1 | |||
XM_011513565.2 | 2059 | Missense Mutation | CGG,TGG | R,W 679 | XP_011511867.1 | |
XM_011513566.1 | 2059 | Missense Mutation | CGG,TGG | R,W 598 | XP_011511868.1 |