Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACTCCCAGGAGGGTGATGGTGA[A/G]CACTATTATTCCACACACCAGGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611124 | ||||||||||||||||||||
Literature Links: |
MFSD8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MFSD8 - major facilitator superfamily domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152778.2 | 2140 | Missense Mutation | CTC,TTC | L,F 494 | NP_689991.1 | |
XM_005262893.1 | 2140 | Missense Mutation | CTC,TTC | L,F 494 | XP_005262950.1 | |
XM_005262896.1 | 2140 | Missense Mutation | CTC,TTC | L,F 445 | XP_005262953.1 | |
XM_005262897.2 | 2140 | Missense Mutation | CTC,TTC | L,F 427 | XP_005262954.1 | |
XM_005262898.2 | 2140 | UTR 3 | XP_005262955.1 | |||
XM_011531830.1 | 2140 | Missense Mutation | CTC,TTC | L,F 456 | XP_011530132.1 | |
XM_011531831.1 | 2140 | Missense Mutation | CTC,TTC | L,F 389 | XP_011530133.1 | |
XM_011531832.1 | 2140 | UTR 3 | XP_011530134.1 | |||
XM_017007989.1 | 2140 | UTR 3 | XP_016863478.1 |