Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTTCTCGCTTGTTCACAGTTTA[C/T]TGCTCTGCTCTGTTTGTTCTGCCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM33 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM33 - transmembrane protein 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018126.2 | 260 | Silent Mutation | TAC,TAT | Y,Y 36 | NP_060596.2 | |
XM_005248116.3 | 260 | Silent Mutation | TAC,TAT | Y,Y 36 | XP_005248173.1 | |
XM_005248117.2 | 260 | Silent Mutation | TAC,TAT | Y,Y 36 | XP_005248174.1 |