Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACTCAGCGTCGTAGAACCTCAG[C/G]CTGTATTTGAGGCATCAGGTGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600571 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
REST PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
REST - RE1 silencing transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193508.1 | 675 | Missense Mutation | CCT,GCT | P,A 123 | NP_001180437.1 | |
NM_005612.4 | 675 | Missense Mutation | CCT,GCT | P,A 123 | NP_005603.3 | |
XM_011534401.2 | 675 | Missense Mutation | CCT,GCT | P,A 123 | XP_011532703.1 | |
XM_017008527.1 | 675 | Missense Mutation | CCT,GCT | P,A 123 | XP_016864016.1 |