Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGTAAGATGGTGAGGGAGTCTG[A/G]ACTTGTGTCTCCATTTGGGATACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIML2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIML2 - tripartite motif family like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303419.1 | 1549 | Missense Mutation | CCA,TCA | P,S 444 | NP_001290348.1 | |
NM_173553.2 | 1549 | UTR 3 | NP_775824.2 | |||
XM_011531737.2 | 1549 | Missense Mutation | CCA,TCA | P,S 419 | XP_011530039.1 | |
XM_011531738.1 | 1549 | Missense Mutation | CCA,TCA | P,S 419 | XP_011530040.1 | |
XM_011531739.1 | 1549 | Missense Mutation | CCA,TCA | P,S 327 | XP_011530041.1 | |
XM_017007882.1 | 1549 | Missense Mutation | CCA,TCA | P,S 419 | XP_016863371.1 |