Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCCCGCCAAACATGAAATGTTC[A/G]CCTTCTCCCTTCCAATGTGATGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608041 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANTXR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANTXR2 - anthrax toxin receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145794.1 | 3488 | Intron | NP_001139266.1 | |||
NM_001286780.1 | 3488 | UTR 3 | NP_001273709.1 | |||
NM_001286781.1 | 3488 | UTR 3 | NP_001273710.1 | |||
NM_058172.5 | 3488 | UTR 3 | NP_477520.2 | |||
XM_011531587.2 | 3488 | Intron | XP_011529889.1 | |||
XM_017007711.1 | 3488 | Intron | XP_016863200.1 |