Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTAAATGGTCCTGGTGAAAAC[A/G]TCAGTGAGGATGAGGCTCAGTCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LARP1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LARP1B - La ribonucleoprotein domain family member 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278604.1 | 1180 | Missense Mutation | ATC,GTC | I,V 59 | NP_001265533.1 | |
NM_018078.3 | 1180 | Missense Mutation | ATC,GTC | I,V 59 | NP_060548.2 | |
NM_032239.3 | 1180 | Missense Mutation | ATC,GTC | I,V 59 | NP_115615.2 | |
NM_178043.2 | 1180 | Missense Mutation | ATC,GTC | I,V 59 | NP_835144.1 | |
XM_011532057.2 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_011530359.2 | |
XM_011532058.2 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_011530360.2 | |
XM_011532059.2 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_011530361.2 | |
XM_011532060.2 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_011530362.2 | |
XM_011532061.2 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_011530363.1 | |
XM_011532062.2 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_011530364.2 | |
XM_011532063.2 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_011530365.2 | |
XM_011532064.2 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_011530366.2 | |
XM_011532065.2 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_011530367.1 | |
XM_011532067.1 | 1180 | Missense Mutation | ATC,GTC | I,V 59 | XP_011530369.1 | |
XM_011532068.1 | 1180 | Missense Mutation | ATC,GTC | I,V 59 | XP_011530370.1 | |
XM_011532069.1 | 1180 | Missense Mutation | ATC,GTC | I,V 59 | XP_011530371.1 | |
XM_011532070.2 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_011530372.2 | |
XM_011532072.2 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_011530374.2 | |
XM_011532073.2 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_011530375.2 | |
XM_011532075.1 | 1180 | UTR 5 | XP_011530377.1 | |||
XM_011532076.2 | 1180 | UTR 5 | XP_011530378.1 | |||
XM_011532077.2 | 1180 | Intron | XP_011530379.1 | |||
XM_017008338.1 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_016863827.1 | |
XM_017008339.1 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_016863828.1 | |
XM_017008340.1 | 1180 | Missense Mutation | ATC,GTC | I,V 59 | XP_016863829.1 | |
XM_017008341.1 | 1180 | Missense Mutation | ATC,GTC | I,V 59 | XP_016863830.1 | |
XM_017008342.1 | 1180 | Missense Mutation | ATC,GTC | I,V 46 | XP_016863831.1 | |
XM_017008343.1 | 1180 | Missense Mutation | ATC,GTC | I,V 59 | XP_016863832.1 | |
XM_017008344.1 | 1180 | Missense Mutation | ATC,GTC | I,V 59 | XP_016863833.1 | |
XM_017008345.1 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_016863834.1 | |
XM_017008346.1 | 1180 | UTR 5 | XP_016863835.1 | |||
XM_017008347.1 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_016863836.1 | |
XM_017008348.1 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_016863837.1 | |
XM_017008349.1 | 1180 | UTR 5 | XP_016863838.1 | |||
XM_017008350.1 | 1180 | Intron | XP_016863839.1 | |||
XM_017008351.1 | 1180 | UTR 5 | XP_016863840.1 | |||
XM_017008352.1 | 1180 | Missense Mutation | ATC,GTC | I,V 260 | XP_016863841.1 | |
XM_017008353.1 | 1180 | Missense Mutation | ATC,GTC | I,V 59 | XP_016863842.1 |