Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGGACACCCAGAGTGGGGAGAGA[C/T]AGTGGCAATGGACGGGCAAAAGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605173 MIM: 606873 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENC1 - ectodermal-neural cortex 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256574.1 | 172 | UTR 5 | NP_001243503.1 | |||
NM_001256575.1 | 172 | UTR 5 | NP_001243504.1 | |||
NM_001256576.1 | 172 | UTR 5 | NP_001243505.1 | |||
NM_003633.3 | 172 | UTR 5 | NP_003624.1 | |||
XM_011543696.2 | 172 | UTR 5 | XP_011541998.1 | |||
XM_011543697.2 | 172 | UTR 5 | XP_011541999.1 |
HEXB - hexosaminidase subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000521.3 | 172 | Intron | NP_000512.1 | |||
NM_001292004.1 | 172 | Intron | NP_001278933.1 |