Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGGGACAGACTGGCCTGGGGAA[C/T]ATTGAAGAGCTTGGAAAGCTGGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609963 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHSY3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHSY3 - chondroitin sulfate synthase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175856.4 | 1253 | Silent Mutation | AAC,AAT | N,N 293 | NP_787052.3 | |
XM_005271982.3 | 1253 | Silent Mutation | AAC,AAT | N,N 293 | XP_005272039.1 | |
XM_005271983.3 | 1253 | Silent Mutation | AAC,AAT | N,N 293 | XP_005272040.1 | |
XM_011543363.2 | 1253 | UTR 5 | XP_011541665.1 | |||
XM_011543364.2 | 1253 | Intron | XP_011541666.1 | |||
XM_011543365.2 | 1253 | Silent Mutation | AAC,AAT | N,N 293 | XP_011541667.1 | |
XM_017009434.1 | 1253 | Intron | XP_016864923.1 | |||
XM_017009435.1 | 1253 | Silent Mutation | AAC,AAT | N,N 293 | XP_016864924.1 |