Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTCCACTTTCTCTGCAGCCCTC[C/T]AGGGGACCCCTCTCTCAGCCACCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606470 | ||||||||||||||||||||
Literature Links: |
NHP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NHP2 - NHP2 ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RMND5B - required for meiotic nuclear division 5 homolog B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288794.1 | 391 | Nonsense Mutation | CAG,TAG | Q,* 49 | NP_001275723.1 | |
NM_001288795.1 | 391 | Nonsense Mutation | CAG,TAG | Q,* 36 | NP_001275724.1 | |
NM_022762.4 | 391 | Nonsense Mutation | CAG,TAG | Q,* 49 | NP_073599.2 | |
XM_005265969.3 | 391 | Nonsense Mutation | CAG,TAG | Q,* 49 | XP_005266026.1 | |
XM_005265971.4 | 391 | UTR 5 | XP_005266028.1 | |||
XM_005265972.3 | 391 | UTR 5 | XP_005266029.1 | |||
XM_017009727.1 | 391 | Nonsense Mutation | CAG,TAG | Q,* 49 | XP_016865216.1 | |
XM_017009728.1 | 391 | UTR 5 | XP_016865217.1 |