Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGGTTCTGACTACATCACCTAT[A/G]GGAATGAATGTCACTTGTGTACCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613511 | ||||||||||||||||||||
Literature Links: |
LOC102546294 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC102546294 - uncharacterized LOC102546294 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPINK7 - serine peptidase inhibitor, Kazal type 7 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032566.2 | 238 | Missense Mutation | AGG,GGG | R,G 61 | NP_115955.1 |
SPINK9 - serine peptidase inhibitor, Kazal type 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |