Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATTTAAATACTGTCTTAGATA[C/T]CTAAAAATTACATTTAAAAAATTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM81B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM81B - family with sequence similarity 81 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152548.2 | Intron | NP_689761.2 | ||||
XM_005271905.2 | Intron | XP_005271962.2 | ||||
XM_005271906.4 | Intron | XP_005271963.2 | ||||
XM_005271907.3 | Intron | XP_005271964.1 | ||||
XM_011543207.1 | Intron | XP_011541509.1 | ||||
XM_011543208.1 | Intron | XP_011541510.1 | ||||
XM_011543209.1 | Intron | XP_011541511.1 | ||||
XM_011543210.2 | Intron | XP_011541512.1 | ||||
XM_017009118.1 | Intron | XP_016864607.1 |