Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTAATAAAAATATTTAAAAACGT[A/G]TCTCAACTTGCATCAATCTTATCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607332 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NREP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NREP - neuronal regeneration related protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142474.1 | Intron | NP_001135946.1 | ||||
NM_001142475.1 | Intron | NP_001135947.1 | ||||
NM_001142476.1 | Intron | NP_001135948.1 | ||||
NM_001142477.1 | Intron | NP_001135949.1 | ||||
NM_001142478.1 | Intron | NP_001135950.1 | ||||
NM_001142479.1 | Intron | NP_001135951.1 | ||||
NM_001142480.1 | Intron | NP_001135952.1 | ||||
NM_001142481.1 | Intron | NP_001135953.1 | ||||
NM_001142482.1 | Intron | NP_001135954.1 | ||||
NM_001142483.1 | Intron | NP_001135955.1 | ||||
NM_004772.2 | Intron | NP_004763.1 | ||||
XM_006714732.1 | Intron | XP_006714795.1 | ||||
XM_006714733.2 | Intron | XP_006714796.1 |
STARD4-AS1 - STARD4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |