Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTAAAGATTCCTCTCCAGTATGG[A/G]TTCTTTTATGCTTGGTGAGATTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610281 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP62 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFP62 - ZFP62 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172638.1 | 2752 | Missense Mutation | ACC,ATC | T,I 861 | NP_001166109.1 | |
NM_152283.4 | 2752 | Missense Mutation | ACC,ATC | T,I 828 | NP_689496.4 | |
XM_017009710.1 | 2752 | Missense Mutation | ACC,ATC | T,I 828 | XP_016865199.1 | |
XM_017009711.1 | 2752 | Missense Mutation | ACC,ATC | T,I 828 | XP_016865200.1 | |
XM_017009712.1 | 2752 | Missense Mutation | ACC,ATC | T,I 828 | XP_016865201.1 | |
XM_017009713.1 | 2752 | Missense Mutation | ACC,ATC | T,I 828 | XP_016865202.1 | |
XM_017009714.1 | 2752 | Missense Mutation | ACC,ATC | T,I 828 | XP_016865203.1 | |
XM_017009715.1 | 2752 | Missense Mutation | ACC,ATC | T,I 828 | XP_016865204.1 | |
XM_017009716.1 | 2752 | Intron | XP_016865205.1 | |||
XM_017009717.1 | 2752 | Missense Mutation | ACC,ATC | T,I 861 | XP_016865206.1 | |
XM_017009718.1 | 2752 | Intron | XP_016865207.1 | |||
XM_017009719.1 | 2752 | Intron | XP_016865208.1 | |||
XM_017009720.1 | 2752 | Intron | XP_016865209.1 | |||
XM_017009721.1 | 2752 | Intron | XP_016865210.1 |