Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCCCAGGCTCCCAGCAGCAGCA[A/G]CAGCAGTGGCAGTGACAGCCTCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609447 MIM: 603790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MZB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MZB1 - marginal zone B and B1 cell specific protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016459.3 | 85 | Silent Mutation | CTG,TTG | L,L 9 | NP_057543.2 |
PROB1 - proline rich basic protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC23A1 - solute carrier family 23 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA24 - spermatogenesis associated 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |